Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chloroaniline-3-sulfonic Acid 98.0+%, TCI America™
SDP

Catalog No. p-7124223 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
25 g
500 g

Chemical Identifiers

CAS 88-43-7
Molecular Formula C6H6ClNO3S
Molecular Weight (g/mol) 207.628
MDL Number MFCD00025296
InChI Key VPXCXBHLKDPWQV-UHFFFAOYSA-N
Synonym 5-Amino-2-chlorobenzenesulfonic Acid
PubChem CID 6933
IUPAC Name 5-amino-2-chlorobenzenesulfonic acid
SMILES C1=CC(=C(C=C1N)S(=O)(=O)O)Cl

Specifications

Quantity 25 g
Formula Weight 207.63
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Chloroaniline-3-sulfonic Acid
EINECSNumber (3)-2019
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.