Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chloroaniline 99.0+%, TCI America™
SDP

Catalog No. p-7124220 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
25 g
500 g

Chemical Identifiers

CAS 106-47-8
Molecular Formula C6H6ClN
Molecular Weight (g/mol) 127.571
MDL Number MFCD00007835
InChI Key QSNSCYSYFYORTR-UHFFFAOYSA-N
Synonym p-chloroaniline, 4-chlorobenzenamine, p-chloraniline, benzenamine, 4-chloro, p-chlorophenylamine, 4-chlorophenylamine, para-chloroaniline, 1-amino-4-chlorobenzene, p-aminochlorobenzene, 4-aminochlorobenzene
PubChem CID 7812
ChEBI CHEBI:20331
IUPAC Name 4-chloroaniline
SMILES C1=CC(=CC=C1N)Cl

Specifications

Melting Point 72°C
Boiling Point 232°C
Quantity 25 g
Physical Form Crystal-Powder at 20°C
UN Number 2018
Formula Weight 127.57
Percent Purity ≥99.0% (GC)
Chemical Name or Material 4-Chloroaniline
EINECSNumber (3)-0194
RTECSNumber BX0700000
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.