Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chlorobenzenesulfonamide 98.0+%, TCI America™
SDP

Catalog No. p-7124792 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
25 g
500 g

Chemical Identifiers

CAS 98-64-6
Molecular Formula C6H6ClNO2S
Molecular Weight (g/mol) 191.63
MDL Number MFCD00007936
InChI Key HHHDJHHNEURCNV-UHFFFAOYSA-N
Synonym p-chlorobenzenesulfonamide, benzenesulfonamide, 4-chloro, p-chlorobenzenesulfamide, 4-chlorophenylsulfonamide, p-chlorophenylsulfonamide, 4-chlorobenzenesulphonamide, benzenesulfonamide, p-chloro, usaf ma-3, 4-chlorobenzene-1-sulfonamide, 4-chloro-benzenesulfonamide
PubChem CID 66824
IUPAC Name 4-chlorobenzene-1-sulfonamide
SMILES NS(=O)(=O)C1=CC=C(Cl)C=C1

Specifications

Melting Point 144°C
Color White
Quantity 25 g
Formula Weight 191.63
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 4-Chlorobenzenesulfonamide
EINECSNumber (3)-1954
RTECSNumber DB1400000
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.