Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4′-Chlorobenzophenone-2-carboxylic Acid 98.0+%, TCI America™
SDP

Catalog No. p-7124778 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
25 g
500 g

Chemical Identifiers

CAS 85-56-3
Molecular Formula C14H9ClO3
Molecular Weight (g/mol) 260.673
MDL Number MFCD00002474
InChI Key YWECCEXWKFHHQJ-UHFFFAOYSA-N
Synonym 2-4-chlorobenzoyl benzoic acid, benzoic acid, 2-4-chlorobenzoyl, o-p-chlorobenzoyl benzoic acid, 2-p-chlorobenzoyl benzoic acid, o-4-chlorobenzoyl benzoic acid, 2-4'-chlorobenzoyl benzoic acid, unii-3u7w4yoy3r, 3u7w4yoy3r, chembl81936, 2-4-chloro-benzoyl-benzoic acid
PubChem CID 66564
IUPAC Name 2-(4-chlorobenzoyl)benzoic acid
SMILES C1=CC=C(C(=C1)C(=O)C2=CC=C(C=C2)Cl)C(=O)O

Specifications

Melting Point 151°C
Color White
Quantity 25 g
Formula Weight 260.67
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4′-Chlorobenzophenone-2-carboxylic Acid
EINECSNumber (4)-0137
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.