Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chlorophenylsulfonylacetone, 98+%, Thermo Scientific™

Catalog No. p-3598204
Click to view available options
Quantity:
1 g
5 g

Chemical Identifiers

CAS 5000-48-6
Molecular Formula C9H9ClO3S
Molecular Weight (g/mol) 232.68
MDL Number MFCD00018663
InChI Key BRDBHPZILGTBFY-UHFFFAOYSA-N
Synonym 4-chlorophenylsulfonylacetone, 1-4-chlorophenyl sulfonyl acetone, 1-4-chlorophenyl sulfonyl propan-2-one, 4-chlorobenzenesulphonylacetone, p-chlorophenylsulfonylacetone, 1-4-chlorobenzenesulfonyl propan-2-one, 4-chlorophenylsulfonyl acetone, 1-4-chlorophenyl sulfonylpropan-2-one, maybridge1_004685
PubChem CID 521269
IUPAC Name 1-(4-chlorophenyl)sulfonylpropan-2-one
SMILES CC(=O)CS(=O)(=O)C1=CC=C(Cl)C=C1

Specifications

Melting Point 89°C to 91°C
Assay Percent Range 98% min. (GC)
Packaging Glass Bottle
Linear Formula CCl6H4SO2CH2COCH3
Quantity 1 g
Solubility Information Solubility in water: insoluble
Formula Weight 232.69
Percent Purity 98+%
Chemical Name or Material 4-Chlorophenylsulfonylacetone
Hazard Category GHS Signal Word: Warning

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.