Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-CMTB, Tocris Bioscience™

Catalog No. p-4816861 Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

FFA2 agonist; also exhibits positive allosteric modulatory activity

Free fatty acid receptor FFA2 agonist (pEC50 = 6.38) and positive allosteric modulator. Binds at a site distinct from the orthosteric site; modulates the activity of short-chain fatty acids at FFA2 via the FFA2 second extracellular loop (ECL2).

Chemical Identifiers

CAS 300851-67-6
Molecular Formula C14H15ClN2OS
Molecular Weight (g/mol) 294.797
InChI Key AZYDQCGCBQYFSE-UHFFFAOYSA-N
Synonym 4-cmtb, 2-4-chlorophenyl-3-methyl-n-thiazol-2-yl butanamide, 2-4-chlorophenyl-3-methyl-n-1,3-thiazol-2-yl butanamide, enamine_000061, d0i3eq, 4-cmtb hplc, 4-chloro-?-1-methylethyl-n-2-thiazolylbenzeneacetamide, 4-chloro-alpha-1-methylethyl-n-2-thiazolylbenzeneacetamide
PubChem CID 4307629
IUPAC Name 2-(4-chlorophenyl)-3-methyl-N-(1,3-thiazol-2-yl)butanamide
SMILES CC(C)C(C1=CC=C(C=C1)Cl)C(=O)NC2=NC=CS2

Specifications

Quantity 10 mg
Formula Weight 294.8
Percent Purity >98%
Chemical Name or Material 4-CMTB
Recommended Storage Store at +4°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.