Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Cyano-4′-ethylbiphenyl 98.0+%, TCI America™
SDP

Catalog No. p-7125706 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 58743-75-2
Molecular Formula C15H13N
Molecular Weight (g/mol) 207.276
MDL Number MFCD00799420
InChI Key DLLIPJSMDJCZRF-UHFFFAOYSA-N
Synonym 4-cyano-4'-ethylbiphenyl, 4'-ethyl-1,1'-biphenyl-4-carbonitrile, 4-ethyl-4'-cyanobiphenyl, 4-4-ethylphenyl benzonitrile, 1,1'-biphenyl-4-carbonitrile, 4'-ethyl, 4'-ethylbiphenyl-4-carbonitrile, 4-4-ethylphenyl benzenecarbonitrile, pubchem2521, 4'-ethyl-4-cyanobiphenyl
PubChem CID 93890
IUPAC Name 4-(4-ethylphenyl)benzonitrile
SMILES CCC1=CC=C(C=C1)C2=CC=C(C=C2)C#N

Specifications

Melting Point 77°C
Color White
Boiling Point 205°C
Quantity 5 g
Formula Weight 207.28
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Cyano-4′-ethylbiphenyl
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.