Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Cyano-4′-undecylbiphenyl 98.0+%, TCI America™
SDP

Catalog No. C32771G Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
1 g

Chemical Identifiers

CAS 65860-74-4
Molecular Formula C24H31N
Molecular Weight (g/mol) 333.519
MDL Number MFCD00505991
InChI Key YIJBPYUXIFSTAP-UHFFFAOYSA-N
Synonym 4′C-Undecyl-4-biphenylcarbonitrile, 4′C-Undecyl-[1,1′C-biphenyl]-4-carbonitrile, 11CB
PubChem CID 105224
IUPAC Name 4-(4-undecylphenyl)benzonitrile
SMILES CCCCCCCCCCCC1=CC=C(C=C1)C2=CC=C(C=C2)C#N

Specifications

Melting Point 53°C
Color White
Quantity 1 g
Formula Weight 333.52
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Cyano-4′-undecylbiphenyl
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.