Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Ethoxyphenyl isothiocyanate 97%, Thermo Scientific™

Catalog No. SB01663EA
Click to view available options
Quantity:
5 g
10 g

Chemical Identifiers

CAS 3460-49-9
Molecular Formula C9H9NOS
Molecular Weight (g/mol) 179.237
InChI Key REEKOMRZYJXXNR-UHFFFAOYSA-N
Synonym 4-ethoxyphenyl isothiocyanate, 4-isothiocyanatophenetol, p-ethoxyphenyl isothiocyanate, 1-ethoxy-4-isothiocyanato-benzene, benzene, 1-ethoxy-4-isothiocyanato, 4-ethoxyphenylisothiocyanate, 4-ethoxybenzenisothiocyanate, acmc-20amjc, pubchem9560, p-ethoxyphenylisothiocyanate
PubChem CID 77006
IUPAC Name 1-ethoxy-4-isothiocyanatobenzene
SMILES CCOC1=CC=C(C=C1)N=C=S

Specifications

Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 10 g
Formula Weight 179.24
Percent Purity 97%
Chemical Name or Material 4-Ethoxyphenyl isothiocyanate
Hazard Category GHS Signal Word: Warning
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.