Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Fluoro-2-nitroaniline 98.0+%, TCI America™
SDP

Catalog No. p-7129650 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 364-78-3
Molecular Formula C6H5FN2O2
Molecular Weight (g/mol) 156.12
MDL Number MFCD00007830
InChI Key PUGDHSSOXPHLPT-UHFFFAOYSA-N
Synonym 2-nitro-4-fluoroaniline, 4-fluoro-2-nitrophenylamine, 4-fluoro-2-nitrobenzeneamine, 4-fluoro-2-nitrobenzenamine, benzenamine, 4-fluoro-2-nitro, 2-amino-5-fluoronitrobenzene, 4-fluoro-2-nitro-aniline, 4-fluoro-2-nitro-phenylamine, 4-fluor-2-nitroanilin, pubchem3421
PubChem CID 67769
IUPAC Name 4-fluoro-2-nitroaniline
SMILES NC1=CC=C(F)C=C1[N+]([O-])=O

Specifications

Melting Point 94°C
Quantity 5 g
Formula Weight 156.12
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Fluoro-2-nitroaniline
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.