Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Fluoro-2-nitrobenzoic Acid 98.0+%, TCI America™
SDP

Catalog No. p-7129867 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
1 g
5 g

Chemical Identifiers

CAS 394-01-4
Molecular Formula C7H4FNO4
Molecular Weight (g/mol) 185.11
MDL Number MFCD00024300
InChI Key YLUCXHMYRQUERW-UHFFFAOYSA-N
Synonym 4-fluoro-2-nitrobenzoic acid, 2-nitro-4-fluorobenzoic acid, 4-fluoro-2-nitrobenzoicacid, benzoic acid, 4-fluoro-2-nitro, 4-fluoro-2-nitro-benzoic acid, 4-fluoro-2-nitrobenzoic acid 2-nitro-4-fluorobenzoic acid, pubchem1311, acmc-1aim5, ksc497m7t, 2-carboxy-5-fluoronitrobenzene
PubChem CID 2737416
SMILES C1=CC(=C(C=C1F)[N+](=O)[O-])C(=O)O

Specifications

Melting Point 150°C
Color Yellow
Quantity 1 g
Formula Weight 185.11
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Fluoro-2-nitrobenzoic Acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.