Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
4-Fluoro-3-nitrotoluene 98.0+%, TCI America™
Shop All Tokyo Chemical Industry (TCI) Products

Click to view available options
Quantity:
5 g
25 g
Specifications
Specifications
Color | Yellow |
Boiling Point | 241°C |
Quantity | 5 g |
Formula Weight | 155.13 |
Percent Purity | ≥98.0% (GC) |
Physical Form | Crystalline Lumps |
Chemical Name or Material | 4-Fluoro-3-nitrotoluene |
Safety and Handling
TSCA | No |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction