Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4′-Fluorochalcone 98.0+%, TCI America™
SDP

Catalog No. F042525G Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
25 g

Chemical Identifiers

CAS 399-10-0
Molecular Formula C15H11FO
Molecular Weight (g/mol) 226.25
MDL Number MFCD00017961
InChI Key VKNQSJQWRINEFS-IZZDOVSWSA-N
Synonym 4'-fluorochalcone, e-1-4-fluorophenyl-3-phenylprop-2-en-1-one, 2e-1-4-fluorophenyl-3-phenylprop-2-en-1-one, ccris 2225, unii-15372zh7kp, 1-4-fluorophenyl-3-phenyl-2-propen-1-one, 2-propen-1-one, 1-4-fluorophenyl-3-phenyl, e-1-4-fluorophenyl-3-phenyl-2-propen-1-one, e-1-4-fluorophenyl-3-phenyl-prop-2-en-1-one, 1-4-fluorophenyl-3-phenylprop-2-en-1-one
PubChem CID 5702577
IUPAC Name (2E)-1-(4-fluorophenyl)-3-phenylprop-2-en-1-one
SMILES FC1=CC=C(C=C1)C(=O)\C=C\C1=CC=CC=C1

Specifications

Melting Point 80°C
Color Yellow
Quantity 25 g
Formula Weight 226.25
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 4′-Fluorochalcone
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.