Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(Fmoc-amino)benzoic acid, 97%, Thermo Scientific Chemicals

Catalog No. p-7035119
Click to view available options
Quantity:
250 mg
1 g

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 185116-43-2
Molecular Formula C22H17NO4
Molecular Weight (g/mol) 359.381
MDL Number MFCD00144888
InChI Key VGSYYBSAOANSLR-UHFFFAOYSA-N
Synonym fmoc-4-abz-oh, fmoc-paba-oh, n-fmoc-4-aminobenzoic acid, 4-9h-fluoren-9-ylmethoxycarbonylamino benzoic acid, 4-9h-fluoren-9-yl methoxy carbonyl amino benzoic acid, 4-9h-fluoren-9-ylmethoxy carbonyl amino benzoic acid, ambotzfaa1645, fmoc-p-aminobenzoic acid, 4-fmoc-amino benzoic acid, chembl46378
PubChem CID 2756082
IUPAC Name 4-(9H-fluoren-9-ylmethoxycarbonylamino)benzoic acid
SMILES C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC4=CC=C(C=C4)C(=O)O

Specifications

Quantity 250 mg
Beilstein 7722986
Formula Weight 359.38
Percent Purity 97%
Chemical Name or Material 4-(Fmoc-amino)benzoic acid
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.