Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Hydroxy-1-cyclohexanecarboxylic Acid delta-Lactone 98.0+%, TCI America™
SDP

Catalog No. p-7130901 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 4350-84-9
Molecular Formula C7H10O2
Molecular Weight (g/mol) 126.155
MDL Number MFCD06797111
InChI Key NVORRKXXHHFWDH-UHFFFAOYSA-N
Synonym 3,6-Ethanotetrahydropyran-2-one, 2-Oxabicyclo[2.2.2]octan-3-one
PubChem CID 138195
IUPAC Name 3-oxabicyclo[2.2.2]octan-2-one
SMILES C1CC2CCC1C(=O)O2

Specifications

Melting Point 128°C
Color White
Boiling Point 123°C
Quantity 5 g
Formula Weight 126.16
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Hydroxy-1-cyclohexanecarboxylic Acid delta-Lactone
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.