Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Hydroxy-2′-nitrobiphenyl 97.0+%, TCI America™
SDP

Catalog No. p-7130708 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
1 g
5 g

Chemical Identifiers

CAS 51264-59-6
Molecular Formula C12H9NO3
Molecular Weight (g/mol) 215.208
MDL Number MFCD00143326
InChI Key NRRCMTITSCFYOH-UHFFFAOYSA-N
Synonym 2′C-Nitrobiphenyl-4-ol, 4-(2-Nitrophenyl)phenol
PubChem CID 15541112
IUPAC Name 4-(2-nitrophenyl)phenol
SMILES C1=CC=C(C(=C1)C2=CC=C(C=C2)O)[N+](=O)[O-]

Specifications

Melting Point 115°C
Quantity 1 g
Formula Weight 215.21
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Hydroxy-2′-nitrobiphenyl
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.