Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Hydroxy-3-nitrophenylacetic acid, 99%, Thermo Scientific™

Catalog No. p-146637
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle

Chemical Identifiers

CAS 10463-20-4
Molecular Formula C8H7NO5
Molecular Weight (g/mol) 197.15
InChI Key QBHBHOSRLDPIHG-UHFFFAOYSA-N
Synonym 4-hydroxy-3-nitrophenylacetic acid, 2-4-hydroxy-3-nitrophenyl acetic acid, 4-hydroxy-3-nitrophenyl acetic acid, 3-nitro-4-hydroxyphenylacetic acid, 3-nitro-4-hydroxy phenylacetic acid, benzeneacetic acid, 4-hydroxy-3-nitro, no2hpa, hydroxynitrophenylaceticacid, acmc-2098cj, epitope id:158629
PubChem CID 447364
ChEBI CHEBI:546274
SMILES C1=CC(=C(C=C1CC(=O)O)[N+](=O)[O-])O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 144.0°C to 147.0°C
Color Colorless to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Linear Formula HOC6H3(NO2)CH2CO2H
Quantity 1 g
Beilstein 10,IV,562
Formula Weight 197.15
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 4-Hydroxy-3-nitrophenylacetic acid, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
EINECSNumber 233-948-0

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.