Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(Hydroxymercury)benzoic Acid Sodium Salt, 98%, Thermo Scientific Chemicals

Catalog No. AC412070010
Click to view available options
Quantity:
Packaging:
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS

138-85-2

Molecular Formula

C7H5HgNaO3

Molecular Weight (g/mol)

360.69

MDL Number

MFCD00002527

InChI Key

BERBQUNVJGVZCX-UHFFFAOYSA-M

Synonym

sodium 4-carboxylatophenyl mercury hydrate, sodium hydrate p-mercuribenzoate

PubChem CID

23667696

IUPAC Name

sodium;(4-carboxylatophenyl)mercury;hydrate

SMILES

C1=CC(=CC=C1C(=O)[O-])[Hg].O.[Na+]

Specifications

CAS Min %

97.5

CAS Max %

100.0

Melting Point

>300°C

Color

White

Infrared Spectrum

Authentic

Assay Percent Range

97.5% min. (ex Hg)

Packaging

Glass bottle

Quantity

1 g

Beilstein

16, 969

Solubility Information

Solubility in water: slightly soluble in water

Formula Weight

360.69

Percent Purity

98%

Physical Form

Crystalline Powder

Chemical Name or Material

4-(Hydroxymercury)benzoic acid sodium salt

Hazard Category

GHS Signal Word: Danger

Hazard Statement

GHS H Statement:
Fatal if inhaled.
Very toxic to aquatic life with long lasting effects.
Fatal if swallowed.
May cause damage to organs through prolonged or repeated exposure.
Fatal in contact with skin.

Precautionary Statement

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Remove/Take off immediately all contaminated clothing.
IF ON SKIN: Gently wash with plenty of soap and water.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber

205-340-5

RTECSNumber

OV8150000

TSCA

TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.