Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Hydroxyphenyl benzoate, 98%, Thermo Scientific Chemicals

Catalog No. p-7077575
Click to view available options
Quantity:
5 g
25 g

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2444-19-1
Molecular Formula C13H10O3
Molecular Weight (g/mol) 214.22
MDL Number MFCD00053304
InChI Key JFAXJRJMFOACBO-UHFFFAOYSA-N
Synonym hydroquinone monobenzoate, p-hydroxyphenyl benzoate, unii-zcy7o225cp, 4-benzoyloxyphenol, 1,4-diphenol, 1-benzoate, benzoic acid 4-hydroxyphenyl ester, 4-hydroxyphenyl benzoate, zcy7o225cp, 4-hydroxyphenylbenzoate, 4-hydroxyphenylbenzoat
PubChem CID 75549
IUPAC Name (4-hydroxyphenyl) benzoate
SMILES OC1=CC=C(OC(=O)C2=CC=CC=C2)C=C1

Specifications

Melting Point 164.0°C to 165.0°C
Loss on Drying 0.5% max.
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Quantity 5 g
Formula Weight 214.22
Percent Purity 98%
Chemical Name or Material 4-Hydroxyphenyl benzoate
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee
EINECSNumber 219-479-4

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.