Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(4-Hydroxyphenyl)phosphonic Acid 97.0+%, TCI America™
SDP

Catalog No. p-7131112 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
200 mg
1 g

Chemical Identifiers

CAS 33795-18-5
Molecular Formula C6H7O4P
Molecular Weight (g/mol) 174.092
MDL Number MFCD01112120
InChI Key YIDVLWDHYNWHMH-UHFFFAOYSA-N
PubChem CID 214694
IUPAC Name (4-hydroxyphenyl)phosphonic acid
SMILES C1=CC(=CC=C1O)P(=O)(O)O

Specifications

Melting Point 169°C
Color White-Yellow
Quantity 200 mg
Formula Weight 174.09
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material (4-Hydroxyphenyl)phosphonic Acid
RTECSNumber SZ8566300
TSCA No
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.