Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Hydroxyphthalic acid, 98%, Thermo Scientific Chemicals

Catalog No. p-4258999
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle

CAS: 610-35-5 | C8H6O5 | 182.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 610-35-5
Molecular Formula C8H6O5
Molecular Weight (g/mol) 182.13
InChI Key MWRVRCAFWBBXTL-UHFFFAOYSA-N
Synonym 1,2-benzenedicarboxylic acid, 4-hydroxy, 4-hydroxybenzene-1,2-dicarboxylic acid, 4-hydroxy-1,2-benzenedicarboxylic acid, 3,4-dicarboxyphenol, 4-hydroxy-phthalic acid, acmc-1avol, ksc357m7h, 4-hydroxyphthalic acid, 4-hydroxy-1,2-benzenedicarboxylicacid, 1,2-benzenedicarboxylicacid, 4-hydroxy
PubChem CID 11881
ChEBI CHEBI:27600
IUPAC Name 4-hydroxyphthalic acid
SMILES C1=CC(=C(C=C1O)C(=O)O)C(=O)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 191.0°C to 194.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 5 g
Solubility Information Solubility in water: insoluble.
Formula Weight 182.13
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 4-Hydroxyphthalic acid
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a
EINECSNumber 210-221-6

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.