Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Iodo-3,5-dimethylpyrazole 98.0+%, TCI America™
SDP

Catalog No. p-7131589 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
1 g
5 g

Chemical Identifiers

CAS 2033-45-6
Molecular Formula C5H7IN2
Molecular Weight (g/mol) 222.029
MDL Number MFCD00040247
InChI Key MZZXIXHKDJNBJQ-UHFFFAOYSA-N
Synonym 3,5-dimethyl-4-iodopyrazole, 4-iodo-3,5-dimethylpyrazole, 3,5-dimethyl-4-iodo-1h-pyrazole, pyrazole, 3,5-dimethyl-4-iodo, 3,5-dimethyl-4-lodopyrazole, 1h-pyrazole, 4-iodo-3,5-dimethyl, zlchem 469, acmc-209f8u, 5-23-05-00159 beilstein handbook reference, ksc490o8h
PubChem CID 97115
IUPAC Name 4-iodo-3,5-dimethyl-1H-pyrazole
SMILES CC1=C(C(=NN1)C)I

Specifications

Melting Point 140°C
Color White-Yellow
Quantity 1 g
Formula Weight 222.03
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Iodo-3,5-dimethylpyrazole
RTECSNumber UQ6517000
TSCA No
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.