Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Iodophenylsulfur Pentafluoride 94.0+%, TCI America™
SDP

Catalog No. p-7131547 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
1 g
5 g

Chemical Identifiers

CAS 286947-68-0
Molecular Formula C6H4F5IS
Molecular Weight (g/mol) 330.05
MDL Number MFCD03788514
InChI Key FRYANWYSCROOCU-UHFFFAOYSA-N
PubChem CID 2779199
IUPAC Name 1-iodo-4-(pentafluoro-λ⁶-sulfanyl)benzene
SMILES FS(F)(F)(F)(F)C1=CC=C(I)C=C1

Specifications

Melting Point 36°C
Color Yellow
Boiling Point 52°C
Quantity 1 g
Formula Weight 330.05
Percent Purity ≥94.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Iodophenylsulfur Pentafluoride
TSCA No
Recommended Storage Refrigerator
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.