Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Isopropylcyclohexanecarboxylic Acid (cis- and trans- mixture) 98.0+%, TCI America™
SDP

Catalog No. I08745G Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g

Chemical Identifiers

CAS 62067-45-2
Molecular Formula C10H18O2
Molecular Weight (g/mol) 170.252
MDL Number MFCD01734696
InChI Key YRQKWRUZZCBSIG-UHFFFAOYSA-N
PubChem CID 81526
IUPAC Name 4-propan-2-ylcyclohexane-1-carboxylic acid
SMILES CC(C)C1CCC(CC1)C(=O)O

Specifications

Color Colorless
Boiling Point 134°C
Quantity 5 g
Formula Weight 170.25
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 4-Isopropylcyclohexanecarboxylic Acid (cis- and trans- mixture)
RTECSNumber GU8585000
TSCA No
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.