Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(Methanesulfonamido)phenylboronic acid, 97%, Thermo Scientific Chemicals

Catalog No. p-4813016
Click to view available options
Quantity:
1 g
5 g

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 380430-57-9
Molecular Formula C7H10BNO4S
Molecular Weight (g/mol) 215.03
MDL Number MFCD02179473
InChI Key NDVJJEADFLTFCD-UHFFFAOYSA-N
Synonym 4-methylsulfonylamino phenylboronic acid, 4-methylsulfonamido phenylboronic acid, 4-methanesulfonylaminophenylboronic acid, 4-methylsulfonylaminophenylboronic acid, 4-methanesulfonylamino phenylboronic acid, 4-methylsulfonamido phenyl boronic acid, 4-methanesulfonamidophenyl boronic acid, 4-methanesulfonamido phenyl boronic acid, 4-methylsulphonyl amino benzeneboronic acid, 4-methylsulfonylamino benzeneboronic acid
PubChem CID 2773537
IUPAC Name [4-(methanesulfonamido)phenyl]boronic acid
SMILES CS(=O)(=O)NC1=CC=C(C=C1)B(O)O

Specifications

Melting Point 234°C to 236°C
Assay Percent Range 96% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 215.04
Percent Purity 97%
Chemical Name or Material 4-(Methanesulfonamido)phenylboronic acid
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.