Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methoxyphenyl 2,4,6-Tri-O-benzyl-beta-D-galactopyranoside 98.0+%, TCI America™
SDP

Catalog No. M15921G Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:

Chemical Identifiers

CAS

247027-79-8

Molecular Formula

C34H36O7

Molecular Weight (g/mol)

556.655

MDL Number

MFCD06797134

InChI Key

MKHXZYRDXXUJMR-BWNLSPMZSA-N

PubChem CID

10769142

IUPAC Name

(2S,3R,4S,5R,6R)-2-(4-methoxyphenoxy)-3,5-bis(phenylmethoxy)-6-(phenylmethoxymethyl)oxan-4-ol

SMILES

COC1=CC=C(C=C1)OC2C(C(C(C(O2)COCC3=CC=CC=C3)OCC4=CC=CC=C4)O)OCC5=CC=CC=C5

Specifications

Melting Point

94°C

Color

White-Yellow

Quantity

1 g

Formula Weight

556.66

Percent Purity

≥98.0% (HPLC)

Physical Form

Crystalline Powder

Chemical Name or Material

4-Methoxyphenyl 2,4,6-Tri-O-benzyl-beta-D-galactopyranoside

TSCA

No

Recommended Storage

Freezer

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.