Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methyl-2-nitrobenzonitrile, 97%, Thermo Scientific™

Catalog No. TL00550EA
Click to view available options
:
Amber Glass Bottle; 10g

Specifications

CAS 26830-95-5
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C8H6N2O2
Linear Formula O2NC6H3(CH3)CN
Quantity 10g
MDL Number MFCD00056107
Synonym 2-nitro-p-tolunitrile, 2-nitro-4-toluonitrile, benzonitrile, 4-methyl-2-nitro, 2-nitro-4-methylbenzonitrile, 4-methyl-2-nitro-benzonitrile, 4-methyl-2-nitrobenzenecarbonitrile, pubchem13730, acmc-1cjm6, ksc494k5h, 4-methyl-2-nitro benzonitrile
InChI Key QGBSLPHQCUIZKK-UHFFFAOYSA-N
SMILES CC1=CC=C(C#N)C(=C1)[N+]([O-])=O
IUPAC Name 4-methyl-2-nitrobenzonitrile
Molecular Weight (g/mol) 162.15
PubChem CID 96235
Formula Weight 162.15
Percent Purity 97%
Chemical Name or Material 4-Methyl-2-nitrobenzonitrile
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
Harmful in contact with skin.
Precautionary Statement GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.