Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(4-Methyl-3,4-dihydro-2H-1,4-benzoxazin-7-yl)methylamine, 97%, Thermo Scientific™

Catalog No. p-3629683
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle

Chemical Identifiers

CAS 946409-08-1
Molecular Formula C10H14N2O
Molecular Weight (g/mol) 178.235
MDL Number MFCD11109315
InChI Key PGIOCCIKSFJJMR-UHFFFAOYSA-N
Synonym 4-methyl-3,4-dihydro-2h-1,4-benzoxazin-7-yl methylamine, 4-methyl-2,3-dihydro-1,4-benzoxazin-7-yl methanamine, 4-methyl-3,4-dihydro-2h-benzo b 1,4 oxazin-7-yl methanamine, 1-4-methyl-2,3-dihydro-1,4-benzoxazin-7-yl methanamine, 4-methyl-2h,3h-benzo e 1,4-oxazin-7-yl methylamine, 1-4-methyl-3,4-dihydro-2h-1,4-benzoxazin-7-yl methanamine
PubChem CID 33589445
IUPAC Name (4-methyl-2,3-dihydro-1,4-benzoxazin-7-yl)methanamine
SMILES CN1CCOC2=C1C=CC(=C2)CN

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 178.23
Percent Purity 97%
Chemical Name or Material (4-Methyl-3, 4-dihydro-2H-1, 4-benzoxazin-7-yl)methylamine
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.