Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methyl-3-nitrobenzoic Acid 99.0+%, TCI America™
SDP

Catalog No. p-7132460 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
25 g
500 g

Chemical Identifiers

CAS 96-98-0
Molecular Formula C8H7NO4
Molecular Weight (g/mol) 181.147
MDL Number MFCD00007174
InChI Key BBEWSMNRCUXQRF-UHFFFAOYSA-N
Synonym 4-methyl-3-nitrobenzoic acid, 3-nitro-4-methylbenzoic acid, 3-nitro-p-toluic acid, benzoic acid, 4-methyl-3-nitro, 3-nitro-para-toluic acid, m-nitro-p-toluic acid, 4-methyl-3-nitro-benzoic acid, p-toluic acid, 3-nitro, 3-nitro-4-methyl benzoic acid, benzoic acid,4-methyl-3-nitro
PubChem CID 7319
SMILES CC1=C(C=C(C=C1)C(=O)O)[N+](=O)[O-]

Specifications

Melting Point 190°C
Color Yellow
Quantity 25 g
Formula Weight 181.15
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Methyl-3-nitrobenzoic Acid
EINECSNumber (3)-1510
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.