Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methylthiazole-5-carboxylic acid, 97%, Thermo Scientific™

Catalog No. MO00777DA
Click to view available options
Quantity:
1 g

Chemical Identifiers

CAS 20485-41-0
Molecular Formula C5H5NO2S
Molecular Weight (g/mol) 143.16
MDL Number MFCD00626872
InChI Key ZGWGSEUMABQEMD-UHFFFAOYSA-N
Synonym 4-methylthiazole-5-carboxylic acid, 4-methyl-5-thiazolecarboxylic acid, 5-thiazolecarboxylic acid, 4-methyl, 4-methyl-thiazole-5-carboxylic acid, 4-methyl-5-thiazole carboxylic acid, 4-methylthiazole-5-carboxylicacid, pubchem8416, acmc-209far, 4-27-00-04008 beilstein handbook reference, ksc201q4f
PubChem CID 209805
IUPAC Name 4-methyl-1,3-thiazole-5-carboxylic acid
SMILES CC1=C(SC=N1)C(=O)O

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 143.17
Percent Purity 97%
Chemical Name or Material 4-Methylthiazole-5-carboxylic acid
Hazard Category Warning
Hazard Statement GHS H Statement:
May cause an allergic skin reaction.
Precautionary Statement GHS P Statement:
Wear eye protection/face protection.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.