Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(Morpholine-4-sulfonyl)-benzenesulfonyl chloride, Technical Grade, Thermo Scientific™

Catalog No. p-4510081
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle

Chemical Identifiers

CAS 465514-13-0
Molecular Formula C10H12ClNO5S2
Molecular Weight (g/mol) 325.778
InChI Key ZZSBUDUHESWZJB-UHFFFAOYSA-N
Synonym 4-morpholin-4-ylsulfonyl benzenesulfonyl chloride, 4-morpholine-4-sulfonyl benzene-1-sulfonyl chloride, 4-morpholinosulfonyl benzene-1-sulfonyl chloride, 4-morpholine-4-sulfonyl benzenesulfonyl chloride, 4-morpholine-4-sulfonyl-benzenesulfonyl chloride, 4-morpholine-4-sulfonyl phenylsulfonyl chloride, 4-morpholine-4-sulfonyl-benzenesulfonylchloride
PubChem CID 2779790
IUPAC Name 4-morpholin-4-ylsulfonylbenzenesulfonyl chloride
SMILES C1COCCN1S(=O)(=O)C2=CC=C(C=C2)S(=O)(=O)Cl

Specifications

Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 325.79
Percent Purity Tech.
Grade Technical
Chemical Name or Material 4-(Morpholine-4-sulfonyl)-benzenesulfonyl chloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.