Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-n-Octyloxyphenol 98.0+%, TCI America™
SDP

Catalog No. p-7134204 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 3780-50-5
Molecular Formula C14H22O2
Molecular Weight (g/mol) 222.328
MDL Number MFCD00045779
InChI Key HFRUPPHPJRZOCM-UHFFFAOYSA-N
Synonym 4-octyloxy phenol, 4-n-octyloxy phenol, 4-n-octyloxyphenol, phenol, 4-octyloxy, 4-octyloxyphenol, p-octyloxyphenol, p-octyloxy phenol, 4-n-octoxyphenol, pubchem13211, acmc-209ivj
PubChem CID 77412
IUPAC Name 4-octoxyphenol
SMILES CCCCCCCCOC1=CC=C(C=C1)O

Specifications

Melting Point 60°C
Boiling Point 127°C
Quantity 5 g
Formula Weight 222.33
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-n-Octyloxyphenol
EINECSNumber (3)-3539
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.