Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Nitro-m-cresol 98.0+%, TCI America™
SDP

Catalog No. p-7133764 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
25 g
500 g

Chemical Identifiers

CAS 2581-34-2
Molecular Formula C7H7NO3
Molecular Weight (g/mol) 153.14
MDL Number MFCD00007333
InChI Key PIIZYNQECPTVEO-UHFFFAOYSA-N
Synonym 4-nitro-m-cresol, 4-nitro-3-cresol, 4-nitro-3-methylphenol, 2-nitro-5-hydroxytoluene, 5-hydroxy-2-nitrotoluene, phenol, 3-methyl-4-nitro, 4-nitro-5-methylphenol, m-cresol, 4-nitro, p-nitro-m-cresol, 4-nitro-1-hydroxy-3-methylbenzene
PubChem CID 17412
ChEBI CHEBI:38683
IUPAC Name 3-methyl-4-nitrophenol
SMILES CC1=CC(O)=CC=C1[N+]([O-])=O

Specifications

Melting Point 128°C
Color Yellow
Quantity 25 g
UN Number 2446
Formula Weight 153.14
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Nitro-m-cresol
EINECSNumber (3)-0790
RTECSNumber GP2625000
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.