Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Nitrobenzoyl chloride, ≥99.0% (GC), MilliporeSigma™ Supelco™

Catalog No. 111018990 Shop All MilliporeSigma Supelco Products
Click to view available options
Quantity:
100 g

4-Nitrobenzoyl chloride is a derivatization reagent. It reacts with L-glutamic acid undergoing methylation using formaldehyde, synthesizing N-(4-methylaminobenzoyl)glutaminic acid.

Chemical Identifiers

MDL Number MFCD00007345

Specifications

Melting Point 71°C to 74°C; 71°C to 74°C
CAS 122-04-3
Boiling Point 202°C to 205°C (105 mmHg)
Flash Point 102°C - Closed Cup
Linear Formula O2 NC6H4 COCl
Quantity 100 g
Formula Weight 185.56
Percent Purity ≥99.0% (GC)
Grade For HPLC derivatization
RTECSNumber DM6651000
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.