Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Nitrophenyl beta-D-Xylopyranoside 98.0+%, TCI America™
SDP

Catalog No. N0620100MG Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:

Chemical Identifiers

CAS

2001-96-9

Molecular Formula

C11H13NO7

Molecular Weight (g/mol)

271.225

MDL Number

MFCD00047519

InChI Key

MLJYKRYCCUGBBV-YTWAJWBKSA-N

Synonym

pnpx, p-nitrophenyl-beta-d-xylopyranoside, 4-nitrophenyl beta-d-xyloside, 3-nitrophenyl b-d-xylopyranoside, p-nitrophenyl beta-d-xylopyranoside, 4-nitrophenyl beta-d-xylopyranoside, 4-nitrophenyl-beta-d-xylopyranoside, p-nitrophenyl beta-d-xyloside, 2s,3r,4s,5r-2-4-nitrophenoxy oxane-3,4,5-triol, xyl1-, a-pnp

PubChem CID

91509

IUPAC Name

(2S,3R,4S,5R)-2-(4-nitrophenoxy)oxane-3,4,5-triol

SMILES

C1C(C(C(C(O1)OC2=CC=C(C=C2)[N+](=O)[O-])O)O)O

Specifications

Melting Point

160°C

Color

White-Yellow

Quantity

100 mg

Formula Weight

271.23

Percent Purity

≥98.0% (HPLC)

Physical Form

Crystalline Powder

Chemical Name or Material

4-Nitrophenyl beta-D-Xylopyranoside [Substrate for beta-Xylosidase]

TSCA

No

Recommended Storage

Refrigerator

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.