Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Nitropyrrole-2-carboxylic Acid Hydrate 99.0+%, TCI America™
SDP

Catalog No. p-7133847 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
1 g
5 g

Chemical Identifiers

CAS 5930-93-8
Molecular Formula C5H4N2O4
Molecular Weight (g/mol) 156.097
MDL Number MFCD00059928
InChI Key SNWGRRCTCGASCN-UHFFFAOYSA-N
PubChem CID 7016560
SMILES C1=C(NC=C1[N+](=O)[O-])C(=O)O

Specifications

Melting Point 217°C
Quantity 1 g
Formula Weight 156.10
Percent Purity ≥99.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Nitropyrrole-2-carboxylic Acid Hydrate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.