Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Phenyl-3-thiosemicarbazide 98.0+%, TCI America™
SDP

Catalog No. p-7134508 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
1 g
25 g

Chemical Identifiers

CAS 5351-69-9
Molecular Formula C7H9N3S
Molecular Weight (g/mol) 167.23
MDL Number MFCD00007615
InChI Key KKIGUVBJOHCXSP-UHFFFAOYSA-N
Synonym 4-phenyl-3-thiosemicarbazide, 4-phenylthiosemicarbazide, n-phenylhydrazinecarbothioamide, hydrazinecarbothioamide, n-phenyl, phenyl thiosemicarbazide, semicarbazide, 4-phenyl-3-thio, 4-phenyl thiosemicarbazide, usaf el-45, usaf ek-5426, 3-amino-1-phenylthiourea
PubChem CID 730679
IUPAC Name 1-amino-3-phenylthiourea
SMILES C1=CC=C(C=C1)NC(=S)NN

Specifications

Melting Point 140°C
Color White
Quantity 1 g
UN Number 2811
Formula Weight 167.23
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Phenyl-3-thiosemicarbazide
EINECSNumber (3)-0123&(3)-2204
RTECSNumber VT4025000
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.