Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(tert-Butyl)phenyl isocyanate, 97%, Thermo Scientific™

Catalog No. p-2700903
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle

Chemical Identifiers

CAS 1943-67-5
Molecular Formula C11H13NO
Molecular Weight (g/mol) 175.231
MDL Number MFCD00040718
InChI Key WUWBDQJTQTVBSQ-UHFFFAOYSA-N
Synonym 4-tert-butylphenyl isocyanate, 4-tert-butylphenylisocyanate, benzene, 1-1,1-dimethylethyl-4-isocyanato, 4-tert-butylphenyl-isocyanate, 4-tert-butyl phenyl isocyanate, 1-tert-butyl-4-isocyanato-benzene, benzene,1-1,1-dimethylethyl-4-isocyanato, 4-t-butylphenylisocyanate, acmc-209ey5, 4-tertbutylphenyl isocyanate
PubChem CID 2735695
IUPAC Name 1-tert-butyl-4-isocyanatobenzene
SMILES CC(C)(C)C1=CC=C(C=C1)N=C=O

Specifications

CAS Min % 97.0
Density 0.986g/mL
Assay Percent Range 97%
Packaging Amber Glass Bottle
Linear Formula (CH3)3CC6H4NCO
Quantity 1 g
Formula Weight 175.23
Percent Purity 97%
Chemical Name or Material 4-(Tert-butyl)phenyl isocyanate
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Harmful if inhaled.
Harmful in contact with skin.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.