Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(tert-Butyl)phenyl isothiocyanate, 97%, Thermo Scientific™

Catalog No. TL00746EB
Click to view available options
Quantity:
10 g
25 g

Chemical Identifiers

CAS 19241-24-8
Molecular Formula C11H13NS
Molecular Weight (g/mol) 191.292
MDL Number MFCD00022059
InChI Key OCGNNCBNRBTUCG-UHFFFAOYSA-N
PubChem CID 140503
IUPAC Name 1-tert-butyl-4-isothiocyanatobenzene
SMILES CC(C)(C)C1=CC=C(C=C1)N=C=S

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Linear Formula (CH3)3CC6H4NCS
Quantity 25 g
Synonym 4-tert-butylphenyl isothiocyanate, 4-t-butylphenyl isothiocyanate, benzene,1-1,1-dimethylethyl-4-isothiocyanato, 4-tert-butyl phenyl isothiocyanate, benzene, 1-1,1-dimethylethyl-4-isothiocyanato, 4-tert-butylphenylisothiocyanate, zlchem 964, 4-t-butylphenylisothiocyanate, p-t-butylphenyl isothiocyanate, 4-tert-butylphenyl-isothiocyanate
Formula Weight 191.3
Percent Purity 97%
Chemical Name or Material 4-(tert-Butyl)phenyl isothiocyanate
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if inhaled.
Harmful in contact with skin.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.