Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

[4-(tert-Butyl)phenyl]methanesulfonyl chloride, Tech., Thermo Scientific™

Catalog No. MO00924DA
Click to view available options
Quantity:
1 g

Chemical Identifiers

CAS 519056-61-2
Molecular Formula C11H15ClO2S
Molecular Weight (g/mol) 246.749
MDL Number MFCD04115413
InChI Key NEJDUPGGBSPJLG-UHFFFAOYSA-N
Synonym 4-tert-butylphenyl methanesulfonyl chloride, 4-tert-butyl phenyl methanesulfonyl chloride, 4-tert-butyl phenyl methanesulfonylchloride, 4-tert-butylphenyl methanesulphonyl chloride, 4-1-butyl phenylmethanesulfonyl chloride, 4-tert-butyl phenyl methyl chlorosulfone
PubChem CID 2794645
IUPAC Name (4-tert-butylphenyl)methanesulfonyl chloride
SMILES CC(C)(C)C1=CC=C(C=C1)CS(=O)(=O)Cl

Specifications

Quantity 1 g
Formula Weight 246.76
Percent Purity Tech.
Grade Technical
Chemical Name or Material [4-(tert-Butyl)phenyl]methanesulfonyl chloride
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.