Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-tert-Butylbenzoic Acid 99.0+%, TCI America™
SDP

Catalog No. p-7118634 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
25 g
500 g

Chemical Identifiers

CAS 98-73-7
Molecular Formula C11H14O2
Molecular Weight (g/mol) 178.23
MDL Number MFCD00002563
InChI Key KDVYCTOWXSLNNI-UHFFFAOYSA-N
Synonym 4-tert-butyl benzoic acid, tbba, p-tert-butylbenzoic acid, benzoic acid, 4-1,1-dimethylethyl, 4-t-butylbenzoic acid, benzoic acid, p-tert-butyl, 4-tert-butyl-benzoic acid, p-t-butylbenzoic acid, 4-tert-butylbenzoicacid, unii-43z7t3vn0r
PubChem CID 7403
ChEBI CHEBI:34443
IUPAC Name 4-tert-butylbenzoic acid
SMILES CC(C)(C)C1=CC=C(C=C1)C(O)=O

Specifications

Melting Point 167°C
Color White
Quantity 25 g
UN Number 3077
Formula Weight 178.23
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-tert-Butylbenzoic Acid
EINECSNumber (3)-1338
RTECSNumber DG4708000
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.