Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
4-tert-Butylbenzylamine 98.0+%, TCI America™
Shop All Tokyo Chemical Industry (TCI) Products

Click to view available options
Quantity:
5 mL
25 mL
Specifications
Specifications
Color | Colorless |
Boiling Point | 124°C |
Quantity | 5 mL |
UN Number | 2735 |
Formula Weight | 163.26 |
Percent Purity | ≥98.0% (GC,T) |
Physical Form | Liquid |
Chemical Name or Material | 4-tert-Butylbenzylamine |
Safety and Handling
EINECSNumber | (3)-4120 |
TSCA | No |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction