Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(Trifluoromethyl)aniline, ≥97%, Thermo Scientific™

Catalog No. SB00719EB
Click to view available options
Quantity:
25 g

Chemical Identifiers

CAS 455-14-1
Molecular Formula C7H6F3N
Molecular Weight (g/mol) 161.127
MDL Number MFCD00064396
InChI Key ODGIMMLDVSWADK-UHFFFAOYSA-N
Synonym 4-trifluoromethyl aniline, 4-aminobenzotrifluoride, p-aminobenzotrifluoride, p-trifluoromethylaniline, benzenamine, 4-trifluoromethyl, 4-trifluoromethyl benzenamine, p-trifluoromethyl aniline, aniline, p-trifluoromethyl, alpha,alpha,alpha-trifluoro-p-toluidine, a,a,a-trifluoro-p-toluidine
PubChem CID 9964
ChEBI CHEBI:40750
IUPAC Name 4-(trifluoromethyl)aniline
SMILES C1=CC(=CC=C1C(F)(F)F)N

Specifications

CAS Min % 97.0
Assay Percent Range 97+%
Linear Formula H2NC6H4CF3
Quantity 25 g
Formula Weight 161.13
Percent Purity 97%
Chemical Name or Material 4-(Trifluoromethyl)aniline
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
May cause damage to organs through prolonged or repeated exposure.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.