Learn More
National Safety Apparel™ 40 Cal ArcGuard™ RevoLite Arc Flash Kit with Lift Front Hood - No Gloves
SDP
Arc flash kit with lift front hood
$2840.00 - $3300.00
Specifications
Ankle Style Open
Arc Flash Rating 40 cal/cm²
Certifications/Compliance CAT 4, NFPA 70 E, CSA Z462, ASTM F1506, OSHA 1910.269
Closure Type FR Hook and Loop
Color Olive Green
View More Specs
Products 6
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
17-001-1934
SDP
View Documents
National Safety Apparel™
KIT4SCLT40NGLFSM
Small
Each for $2,980.00
missing translation for 'QuantityLeft'
Add to Cart
17-001-1935
SDP
View Documents
National Safety Apparel™
KIT4SCLT40NGLFMD
Medium
Each for $3,190.00
missing translation for 'QuantityLeft'
Add to Cart
17-001-1936
SDP
View Documents
National Safety Apparel™
KIT4SCLT40NGLFLG
Large
Each for $3,250.00
missing translation for 'QuantityLeft'
Add to Cart
17-001-1937
SDP
View Documents
National Safety Apparel™
KIT4SCLT40NGLFXL
X-Large
Each for $3,180.00
missing translation for 'QuantityLeft'
Add to Cart
17-001-1938
SDP
View Documents
National Safety Apparel™
KIT4SCLT40NGLF2X
2X-Large
Each for $3,300.00
missing translation for 'QuantityLeft'
Add to Cart
17-001-1939
SDP
View Documents
National Safety Apparel™
KIT4SCLT40NGLF3X
3X-Large
Each for $2,840.00
missing translation for 'QuantityLeft'
Add to Cart
Description
Description
  • Arc Rating: 40 cal/cm²
  • CAT 4
  • Fabric/Material: 11.2 oz. RevoLite™
  • 50% lighter than our standard 40 cal kits
  • FR hook and loop front closure
  • Stand up collar for extra coverage
  • Rib knit cuffs for comfortable, secure fit
  • Garments embroidered with arc rating
  • Quick disconnect and adjustable shoulder straps on bib overalls
  • Expandable leg opening to fit over boots
  • Made in the USA
Specifications
Specifications
Open
CAT 4, NFPA 70 E, CSA Z462, ASTM F1506, OSHA 1910.269
Olive Green
Stitched
40 cal/cm²
FR Hook and Loop
RevoLite™
Adjustable
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.