Learn More
Ted Pella Inc 40 NM GOLD COLLOIDS
Supplier: Ted Pella Inc 1570720
Gold Colloids, 40nm, 9.0 x 1010, 20mL, Room Temperature
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.