Learn More
Agilent BioTek 406 FX Product Qualification Package
406 FX Product Qualification Package for use with the 406 FX Washer Dispenser.
Supplier: Agilent BioTek 2120530
Description
- Includes thorough procedures to allow a user to perform Installation Qualification, Operational Qualification and Performance Qualification
- Periodic maintenance procedures are also included to assist in maintaining the instruments for optimal performance
- Product Qualification Packages contain only documentation and procedures
- Many of these procedures require the use of QC Test Solutions or other accessories, and these are ordered separately
Specifications
Service Kit | |
Agilent BioTek 406 FX Washer Dispenser |