Learn More
Simco 44IN PLSD / STDY STATE STNDLN
Supplier:  Simco 915225SC44R
Encompass
44" Pulsed & Steady State Standalone AeroBar (24VAC) with FMS Alarm Output and Titanium Emitter Points (9), RoHS. Requires Handheld Remote Below (91-5570) (No Interface Module Required)
Catalog No. NC3353856
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.