Learn More
Ace Glass, Inc. 45/50 NO.25 DISP TUBE ADAP
Supplier: Ace Glass, Inc. 684611
Dispersion tube adapter for insertion between the condenser and extraction body in lighter than water extraction. Features 45/50 joints and a #25 Ace-Thred capillary tip.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.