Learn More
Ace Glass, Inc. 455MM 24/40 4PRT MANIFOLD
Supplier:  Ace Glass, Inc. 873938
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
455mm long double tube vacuum manifold with four #15 o-ring joint ends. Manifold has four stopcock ports with 24/40 inner joints and 0-10MM Easy Action stopcocks. Distance between ports is 100MM which will allow for flasks up to 250mL. 
Catalog No. 50-976-291
missing translation for 'QuantityLeft'