Learn More
Brady™ SPC MAXX Enhanced Oil Only Absorbent Pads
Catalog No. p-4955323 Shop All Brady Corporation Products
Click to view available options
Weight Capacity:
Capacity (English):
Dimensions (L x W):
Made with MAXX Technology for use with oil and petroleum based fluids
  • Hydrophobic and hence ideal for outdoor applications
  • Heavier and bonded for more strength and reduce linting
  • Dimpled and perforated, single-ply sorbents
  • Available in various sizes
Applications:
For Overspray Leaks and Drips, Workstations, Drums - Liquid Storage, Outfalls and Discharge Ponds
Specifications
Capacity (English)
40 gal./Bale
Type
Absorbent Pad, Oil-only
For Use With (Application)
Overspray leaks and drips, workstations, drums - liquid storage, outfalls and discharge ponds
Packaging Quantity
200/Cs.
Weight Capacity
Lightweight
Dimensions (L x W)
38.1 x 48.2 cm (15 x 19 in.)